C12H580D8868 Chemcia Scientific, LLC

5-Bromo-3-chloro-1H-pyrazolo[3,4-c]pyridine

  • MDL number MFCD13177849
  • Cas Number : 929617-31-2
  • Molecular Weight : 232.46791

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0-5*C
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=7384

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7384
Availability details for
Quantity
  • Availability
  • Add to Cart