C12H580D8869 Chemcia Scientific, LLC

5-Bromo-3-chloro-2H-pyrazolo[3,4-c]pyridine

  • MDL number MFCD30180208
  • Molecular Weight : 232.46791

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0-5*C
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=7385

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7385
Availability details for
Quantity
  • Availability
  • Add to Cart