C12H580D886C Chemcia Scientific, LLC

5-Chloro-1H-pyrazolo[3,4-c]pyridin-3-ylamine

  • MDL number MFCD13177857
  • Cas Number : 245325-32-0
  • Molecular Weight : 168.58655

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0-5*C
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=7405

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7405
Availability details for
Quantity
  • Availability
  • Add to Cart