C12H580D886D Chemcia Scientific, LLC

5-Chloro-2H-pyrazolo[3,4-c]pyridin-3-ylamine

  • MDL number MFCD28159363
  • Cas Number : 245325-32-0
  • Molecular Weight : 168.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0-5C
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=7406

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7406
Availability details for
Quantity
  • Availability
  • Add to Cart