C12H580D8870 Chemcia Scientific, LLC

4-Bromo-5-chloro-2-methoxy-benzoic acid methyl ester

  • MDL number MFCD16873052
  • Cas Number : 1239743-42-0
  • Molecular Weight : 279.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0-5C
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=7568

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7568
Availability details for
Quantity
  • Availability
  • Add to Cart