C12H580D8875 Chemcia Scientific, LLC

1-(2-Amino-4-fluoro-5-methoxy-phenyl)-ethanone

  • MDL number MFCD11113349
  • Cas Number : 949159-97-1
  • Molecular Weight : 183.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0-5C
Physical Form Grey Green Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=8126

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=8126
Availability details for
Quantity
  • Availability
  • Add to Cart