C12H580D8876 Chemcia Scientific, LLC

1-(4-Fluoro-5-methoxy-2-nitro-phenyl)-ethanone

  • MDL number MFCD11977400
  • Cas Number : 949159-96-0
  • Molecular Weight : 213.16681

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. 0-5*C
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=8127

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=8127
Availability details for
Quantity
  • Availability
  • Add to Cart