C12H580D887E Chemcia Scientific, LLC

1,4-Dichloro-2-fluoro-5-nitro-benzene

  • MDL number MFCD25961694
  • Cas Number : 2624-66-0
  • Molecular Weight : 209.99274

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0-5*C
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=8799

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=8799
Availability details for
Quantity
  • Availability
  • Add to Cart