C12H580D8882 Chemcia Scientific, LLC

4-Fluoro-1H-pyrazolo[3,4-c]pyridine

  • MDL number MFCD28501792
  • Cas Number : 1373168-65-0
  • Molecular Weight : 137.11728

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0-5*C
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=9156

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9156
Availability details for
Quantity
  • Availability
  • Add to Cart