C12H580D8887 Chemcia Scientific, LLC

5-Bromo-4-fluoro-2-iodo-phenylamine

  • MDL number MFCD30834158
  • Cas Number : 1265236-37-0
  • Molecular Weight : 315.91228

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0-5*C
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=9395

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9395
Availability details for
Quantity
  • Availability
  • Add to Cart