C12H580D8888 Chemcia Scientific, LLC

2-Fluoro-4-methoxy-5-nitro-benzaldehyde

  • MDL number MFCD27995599
  • Cas Number : 586412-86-4
  • Molecular Weight : 199.13972

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0-5*C
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=9539

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9539
Availability details for
Quantity
  • Availability
  • Add to Cart