C12H580D888A Chemcia Scientific, LLC

5-Chloro-2-methoxy-4-methyl-benzoic acid

  • MDL number MFCD14537196
  • Cas Number : 28478-41-3
  • Molecular Weight : 200.62328

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0-5*C
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=9573

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9573
Availability details for
Quantity
  • Availability
  • Add to Cart