C12H580D888B Chemcia Scientific, LLC

3-Bromo-[1,2,4]triazolo[4,3-a]pyridin-6-ylamine

  • MDL number MFCD17677031
  • Cas Number : 1263283-48-2
  • Molecular Weight : 213.03755

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0-5*C
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=9815

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9815
Availability details for
Quantity
  • Availability
  • Add to Cart