C12H580D8892 Chemcia Scientific, LLC

2-Amino-5-fluoro-4-nitro-phenol

  • Cas Number : 1850312-96-7
  • Molecular Weight : 172.11675

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0-5*C
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=10716

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10716
Availability details for
Quantity
  • Availability
  • Add to Cart