C12H580D8894 Chemcia Scientific, LLC

1H-Pyrido[3,4-b][1,4]thiazin-2-one

  • MDL number MFCD28605613
  • Cas Number : 1451041-77-2
  • Molecular Weight : 166.20267

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0-5*C
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=10857

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10857
Availability details for
Quantity
  • Availability
  • Add to Cart