C12H580D889A Chemcia Scientific, LLC

5-Chloro-7-nitro-1H-indole-2,3-dione

  • Cas Number : 153255-76-6
  • Molecular Weight : 226.57711

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0-5*C
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=11876

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=11876
Availability details for
Quantity
  • Availability
  • Add to Cart