C12H580D88A3 Chemcia Scientific, LLC

6-Methoxy-5-nitro-2H-indazole

  • Molecular Weight : 193.16329

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0-5*C
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=12151

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=12151
Availability details for
Quantity
  • Availability
  • Add to Cart