C12H580D88A5 Chemcia Scientific, LLC

6-Methoxy-2H-indazol-5-ylamine

  • Molecular Weight : 163.18043

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0-5*C
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=12153

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=12153
Availability details for
Quantity
  • Availability
  • Add to Cart