C12H580D88B0 Chemcia Scientific, LLC

5-Fluoro-6-methyl-pyridine-2-carbonitrile

  • MDL number MFCD18258166
  • Cas Number : 915720-64-8
  • Molecular Weight : 136.1297

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0-5*C
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=13132

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=13132
Availability details for
Quantity
  • Availability
  • Add to Cart