C12H580D88B1 Chemcia Scientific, LLC

5-Fluoro-3-nitro-1H-indole

  • Cas Number : 1613516-83-8
  • Molecular Weight : 180.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0-5C
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=13175

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=13175
Availability details for
Quantity
  • Availability
  • Add to Cart