C12H580D88B3 Chemcia Scientific, LLC

5-Dimethylamino-2-nitro-benzoic acid

  • Cas Number : 62876-66-8
  • Molecular Weight : 210.19105

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0-5*C
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=13493

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=13493
Availability details for
Quantity
  • Availability
  • Add to Cart