C12H580D88B5 Chemcia Scientific, LLC

4,5-Dichloro-2-methyl-pyridine

  • MDL number MFCD09743863
  • Cas Number : 514216-44-5
  • Molecular Weight : 162.01945

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0-5*C
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=13993

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=13993
Availability details for
Quantity
  • Availability
  • Add to Cart