C12H580D88BB Chemcia Scientific, LLC

5-Chloro-2H-pyrazolo[4,3-b]pyridin-3-ylamine

  • Molecular Weight : 168.58655

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0-5*C
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=15286

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=15286
Availability details for
Quantity
  • Availability
  • Add to Cart