C12H580D88C4 Chemcia Scientific, LLC

3-Amino-6-methoxy-pyridine-2-carbonitrile

  • MDL number MFCD07780779
  • Cas Number : 1314902-70-9
  • Molecular Weight : 149.15334

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0-5*C
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=15487

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=15487
Availability details for
Quantity
  • Availability
  • Add to Cart