C12H580D88C5 Chemcia Scientific, LLC

6-Methoxy-3-nitro-pyridine-2-carboxylic acid amide

  • MDL number MFCD07780781
  • Cas Number : 1314960-41-2
  • Molecular Weight : 197.15154

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0-5*C
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=15488

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=15488
Availability details for
Quantity
  • Availability
  • Add to Cart