C12H580D88CA Chemcia Scientific, LLC

2-Methyl-5,7-dihydro-pyrrolo[3,4-d]pyrimidine-6-carboxylic acid tert-butyl ester

  • MDL number MFCD12400787
  • Cas Number : 1160995-19-6
  • Molecular Weight : 235.28819

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0-5*C
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1490

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1490
Availability details for
Quantity
  • Availability
  • Add to Cart