C12H580D88D1 Chemcia Scientific, LLC

5-Hydroxy-2-methyl-4-nitro-benzoic acid methyl ester

  • Molecular Weight : 211.17578

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0-5*C
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=15731

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=15731
Availability details for
Quantity
  • Availability
  • Add to Cart