C12H93E354E1 Chemcia Scientific, LLC

(2-Amino-4-bromo-phenyl)-acetonitrile

  • MDL number MFCD18389427
  • Cas Number : 1261836-91-2
  • Molecular Weight : 211.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0-5C
Physical Form Grey Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=9563

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9563
Availability details for
Quantity
  • Availability
  • Add to Cart