C12H93E354E4 Chemcia Scientific, LLC

6-Bromo-8-methyl-[1,2,4]triazolo[1,5-a]pyridin-2-ylamine

  • MDL number MFCD23381261
  • Cas Number : 1367782-03-3
  • Molecular Weight : 227.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0-5C
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=9751

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9751
Availability details for
Quantity
  • Availability
  • Add to Cart