C12H93E354E8 Chemcia Scientific, LLC

6-Benzyloxy-3-bromo-1H-quinolin-4-one

  • MDL number MFCD30285244
  • Cas Number : 1777589-98-6
  • Molecular Weight : 330.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0-5C
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=9881

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9881
Availability details for
Quantity
  • Availability
  • Add to Cart