C12H93E35522 Chemcia Scientific, LLC

3-Bromo-2H-indazole-5-carbaldehyde

  • Molecular Weight : 225.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form White Solid
Country of Origin CN
Product Link http://www.chemcia.com/product_fl02.asp?id=14315

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=14315
Availability details for
Quantity
  • Availability
  • Add to Cart