C12H93E35527 Chemcia Scientific, LLC

(3-Bromo-4-fluoro-phenyl)-methyl-amine

  • MDL number MFCD11655722
  • Cas Number : 1233521-11-3
  • Molecular Weight : 204.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form Light Yellow Liquid
Country of Origin CN
Product Link http://www.chemcia.com/product_fl02.asp?id=16148

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=16148
Availability details for
Quantity
  • Availability
  • Add to Cart