CH6371376274 Chem Impex International, Inc.

Adenosine-5'-diphosphate trilithium salt trihydrate

  • MDL number MFCD06795763
  • Cas Number : 31008-64-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5'-ADP 3Li
Purity 99
Storage temp. Store at -20
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/adenosine-5-diphosphate-trilithium-salt/11?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/adenosine-5-diphosphate-trilithium-salt/11?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart