CH6371376338 Chem Impex International, Inc.

Cytidine-5'-diphosphate trisodium salt

  • MDL number MFCD00065201
  • Cas Number : 34393-59-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5'-CDP 3Na
Purity 0.99
Storage temp. Store at =-4
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/cytidine-5-diphosphate-trisodium-salt/80?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/cytidine-5-diphosphate-trisodium-salt/80?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart