CH6371376352 Chem Impex International, Inc.

2'-Deoxyguanosine-5'-monophosphate disodium salt

  • MDL number MFCD00080331
  • Cas Number : 33430-61-4
  • Molecular Weight : 391.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point > 245 °C (dec.)
Purity 99
Storage temp. Store at 0 - 8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-deoxyguanosine-5-monophosphate-disodium-salt/95?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-deoxyguanosine-5-monophosphate-disodium-salt/95?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart