CH6371376356 Chem Impex International, Inc.

P1,P5-Di(adenosine-5')pentaphosphate trilithium salt

  • MDL number MFCD00044936
  • Cas Number : 75522-97-3
  • Molecular Weight : 934.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0 - 8
Physical Form White powder or almost white to slightly yellowish lyophilisate
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/pi-sup-1-sup-p-sup-5-sup-di-adenosine-5-pentaphosphate-trilithium-salt/99?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/pi-sup-1-sup-p-sup-5-sup-di-adenosine-5-pentaphosphate-trilithium-salt/99?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart