CH6371376371 Chem Impex International, Inc.

5'-O-(4,4'-Dimethoxytrityl)-N4-benzoyl-5-methyl-2'-deoxycytidine

  • MDL number MFCD00057965
  • Cas Number : 104579-03-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5'-DMT-NBz-5-Methyl dC
Purity 98
Storage temp. Store at 0-8
Physical Form White, off-white to yellow powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/5-i-o-i-4-4-dimethoxytrityl-i-n-i-sup-4-sup-benzoyl-5-methyl-2-deoxycytidine/116?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-i-o-i-4-4-dimethoxytrityl-i-n-i-sup-4-sup-benzoyl-5-methyl-2-deoxycytidine/116?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart