CH6371376374 Chem Impex International, Inc.

2'-Deoxythymidine-5'-triphosphate trisodium salt

  • MDL number MFCD00080338
  • Cas Number : 18423-43-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2'-dTTP trisodium salt
Purity 98
Storage temp. Store at = -20
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-deoxythymidine-5-triphosphate-trisodium-salt/122?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-deoxythymidine-5-triphosphate-trisodium-salt/122?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart