CH6371376468 Chem Impex International, Inc.

Piperazine-N,N'-bis(2-hydroxypropnane-sulfonic acid) dihydrate

  • MDL number MFCD00150549
  • Cas Number : 68189-43-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at RT
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/piperazine-n-n-i-bis-i-2-hydroxypropnane-sulfonic-acid/226?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/piperazine-n-n-i-bis-i-2-hydroxypropnane-sulfonic-acid/226?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart