CH6371376518 Chem Impex International, Inc.

Vancomycin hydrochloride

  • MDL number MFCD03613611
  • Cas Number : 1404-93-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 2 - 8
Physical Form White or almost white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/vancomycin-hydrochloride/282?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/vancomycin-hydrochloride/282?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart