CH6371376539 Chem Impex International, Inc.

Guanosine-5'-triphosphate disodium salt

  • MDL number MFCD00083629
  • Cas Number : 56001-37-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. Store at -20 °C
Physical Form White to off-white powder
Shipping Temperature Ships with blue ice
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/guanosine-5-triphosphate-disodium-salt/308?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/guanosine-5-triphosphate-disodium-salt/308?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart