CH6371376566 Chem Impex International, Inc.

5-Bromo-2'-deoxycytidine

  • MDL number MFCD00047496
  • Cas Number : 1022-79-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8 °C
Physical Form Off-white to faint yellow powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/5-bromo-2-deoxycytidine/340?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-bromo-2-deoxycytidine/340?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart