CH6371376572 Chem Impex International, Inc.

5-Iodo-2'-deoxycytidine

  • MDL number MFCD00038063
  • Cas Number : 611-53-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-IdC
Purity 99
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/5-iodo-2-deoxycytidine/346?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-iodo-2-deoxycytidine/346?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart