CH6371376593 Chem Impex International, Inc.

Boc-O-3-bromobenzyl-3-iodo-L-tyrosine

  • MDL number MFCD00080286

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124-132
Synonym Boc-3-I-L-Tyr(3-BrBzl)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-o-i-3-bromobenzyl-3-iodo-l-tyrosine/371?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-o-i-3-bromobenzyl-3-iodo-l-tyrosine/371?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart