CH6371376668 Chem Impex International, Inc.

Arg-Asp-OH

  • MDL number MFCD00055818
  • Cas Number : 15706-88-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Arginyl-L-aspartic acid
Purity 97
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/arg-asp-oh/459?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/arg-asp-oh/459?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart