CH6371376694 Chem Impex International, Inc.

Uridine-3'-monophosphate disodium salt

  • MDL number MFCD00042791
  • Cas Number : 35170-03-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3'-UMP 2Na
Purity 99
Storage temp. Store at = -4
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/uridine-3-monophosphate-disodium-salt/489?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/uridine-3-monophosphate-disodium-salt/489?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart