CH6371376695 Chem Impex International, Inc.

Adenosine-3'-monophosphate sodium salt

  • MDL number MFCD00065985
  • Cas Number : 4958-39-8
  • Molecular Weight : 391.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/adenosine-3-monophosphate-sodium-salt/490?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/adenosine-3-monophosphate-sodium-salt/490?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart