CH6371376815 Chem Impex International, Inc.

1,2-bis(2-Aminophenoxy)ethane-N,N,N',N'-tetraacetic acid tetrapotassium salt

  • MDL number MFCD00070080
  • Cas Number : 73630-08-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym BAPTA 4K
Purity 98
Storage temp. Store at RT
Physical Form White to off-white crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/1-2-i-bis-i-2-aminophenoxy-ethane-n-n-n-n-tetraacetic-acid-tetrapotassium-salt/627?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/1-2-i-bis-i-2-aminophenoxy-ethane-n-n-n-n-tetraacetic-acid-tetrapotassium-salt/627?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart