CH6371376826 Chem Impex International, Inc.

N2-Isobutyrylguanosine

  • MDL number MFCD00274102
  • Cas Number : 64350-24-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N2-Isobutyryl-D-guanosine
Purity 97
Storage temp. Store at =-15
Physical Form White to pale yellow powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-2-sup-isobutyrylguanosine/638?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-2-sup-isobutyrylguanosine/638?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart