CH6371376828 Chem Impex International, Inc.

5'-O-(4,4'-Dimethoxytrityl)-2'-deoxyuridine

  • MDL number MFCD00063410
  • Cas Number : 23669-79-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5'-O-DMT-2'-deoxyuridine
Purity 99
Storage temp. Store at =-15
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/5-i-o-i-4-4-dimethoxytrityl-2-deoxyuridine/640?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-i-o-i-4-4-dimethoxytrityl-2-deoxyuridine/640?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart